4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(4-Fluoro-3-(trifluoromethyl)phenyl)ethanone

A16501.03 A16501.06
ALFAA16501.03EA 0 SGD
ALFAA16501.03 ALFAA16501.06
4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%
4'-Fluoro-3'-(trifluoromethyl)acetophenone

Purity: 97% CAS No.: 208173-24-4 Molecular Formula: C9H6F4O Formula Weight: 206.14 Refractive Index: 1.4540

Formula: C₉H₆F₄O
MW: 206,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061252
CAS Number: 208173-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR