2-Hydroxyethyl phenyl sulphide ≥99%
Supplier: Thermo Scientific
Synonyms:
2-(Phenylthio)ethanol
A10861.09
A10861.18
ALFAA10861.09EA
0
SGD
ALFAA10861.09
ALFAA10861.18
2-Hydroxyethyl phenyl sulphide ≥99%
2-Hydroxyethyl phenyl sulphide
Purity: 99%
CAS No.: 699-12-7
Molecular Formula: C6H5S(CH2)2OH
Formula Weight: 154.23
Flash Point:
Formula:
C₈H₁₀OS MW: 154,23 g/mol Boiling Pt: 115…116 °C (2 mmHg) Density: 1,147 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002905 CAS Number: 699-12-7 EINECS: 211-828-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...