E-Z 96® Plant DNA kit
Supplier: OMEGA BIO-TEK
The E-Z 96® Plant DNA kit allows rapid and reliable isolation of high quality total cellular DNA from a wide variety of plant species and tissues in a 96-well format. E-Z 96® Plant DNA kits utilise a buffer system adapted from the SP Plant DNA system, which is suitable for a variety of plants, such as those with unusually high levels of phenolic compounds or polysaccharides (for example, cotton, pine and peanut samples). Up to 50 mg of wet tissue or 12 mg dry tissue can be processed in each well in less than 1 hour. The system combines the reversible nucleic acid-binding properties of the HiBind matrix with the speed and versatility of the E-Z 96® DNA plate to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. Purified DNA is suitable for PCR, restriction digestion, and hybridisation techniques.
- DNA isolation of 96 samples in <90 minutes
- Optimised buffers guarantee pure DNA very time
- No organic extractions
- Purified DNA suitable for most downstream applications
- E-Z 96® plate is suitable for most robotic workstations