(Methylthio)acetaldehyde dimethyl acetal ≥97%
Supplier: Thermo Scientific
|
Warning
|
B22667.06
B22667.14
ALFAB22667.06EA
0
SGD
ALFAB22667.06
ALFAB22667.14
(Methylthio)acetaldehyde dimethyl acetal ≥97%
(Methylthio)acetaldehyde dimethyl acetal
Purity: 97%
CAS No.: 40015-15-4
Molecular Formula: CH3SCH2CH(OCH3)2
Formula Weight: 136.21
Flash Point: 53°(127°F)
Refractive Index: 1.4528
Boiling Point: 170-171°
Beilstein Reference: 773689
UN No.: 1993
MDL No.: MFCD00009848
Formula:
C₅H₁₂O₂S MW: 136,21 g/mol Boiling Pt: 170…171 °C Density: 1,022 Flash Pt: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009848 CAS Number: 40015-15-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...