5-Pyrimidineboronic acid ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid

H52984.MD H52984.03
ALFAH52984.MDEA 0 SGD
ALFAH52984.MD ALFAH52984.03
5-Pyrimidineboronic acid ≥98%
5-Pyrimidineboronic acid

Purity: 98% CAS No.: 109299-78-7 Molecular Formula: C4H5BN2O2 Formula Weight: 123.91 Melting Point: 300C MDL No.: MFCD03002366

Formula: C₄H₅BN₂O₂
MW: 123,91 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03002366
CAS Number: 109299-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR