2-Fluoro-5-formylphenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-5-formylbenzeneboronic acid
H28857.06
H28857.03
ALFAH28857.06EA
0
SGD
ALFAH28857.06
ALFAH28857.03
2-Fluoro-5-formylphenylboronic acid ≥97%
2-Fluoro-5-formylphenylboronic acid
Purity: 97%
CAS No.: 352534-79-3
Molecular Formula: C7H6BFO3
Formula Weight: 167.93
Melting Point: 168-171°
MDL No.: MFCD03095100
Formula:
C₇H₆BFO₃ MW: 167,93 g/mol Melting Pt: 168…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095100 CAS Number: 352534-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...