3,4-Dichlorobenzoic anhydride 95%
Supplier: Thermo Scientific
|
Warning
|
H32900.03
H32900.06
ALFAH32900.03EA
0
SGD
ALFAH32900.03
ALFAH32900.06
3,4-Dichlorobenzoic anhydride 95%
3,4-Dichlorobenzoic anhydride
Purity: 95%
CAS No.: 86866-14-0
Molecular Formula: C14H6Cl4O3
Formula Weight: 364.01
Melting Point: 153-158°
MDL No.: MFCD09757545
Formula:
C₁₄H₆Cl₄O₃ MW: 364,01 g/mol Melting Pt: 153…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757545 CAS Number: 86866-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...