Chlorodimethyloctylsilane 97%, rods

Supplier: Thermo Scientific
Danger

L16564.06
ALFAL16564.06EA 77.5 SGD
ALFAL16564.06
Chlorodimethyloctylsilane 97%, rods
Chlorodimethyloctylsilane

Purity: 97% CAS No.: 18162-84-0 Molecular Formula: C10H23ClSi Formula Weight: 206.83 Flash Point: 92°(198°F) Refractive Index: 1.4350 Odour: Acrid Boiling Point: 222-225° Beilstein Reference: 1738376 UN No.: 2987 MDL No.: MFCD00000511

Formula: C₁₀H₂₃ClSi
MW: 206,83 g/mol
Boiling Pt: 222…225 °C
Density: 0,873
Flash Pt: 92 °C (198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000511
CAS Number: 18162-84-0
EINECS: 242-044-5
UN: 2987
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR