3-Amino-5-fluoropyridine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Fluoropyridin-3-amine

H34111.03 H34111.06
ALFAH34111.03EA 0 SGD
ALFAH34111.03 ALFAH34111.06
3-Amino-5-fluoropyridine 98%
3-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
MW: 112,11 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00234073
CAS Number: 210169-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR