2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine 95%

Supplier: Thermo Scientific
Danger

L20009.MC L20009.ME
ALFAL20009.MCEA 0 SGD
ALFAL20009.MC ALFAL20009.ME
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine 95%
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine

Purity: 95% CAS No.: 675602-89-8 Molecular Formula: C6H3F5N2 Formula Weight: 198.10 Melting Point: 66-68° UN No.: 2811 MDL No.: MFCD03840200

Formula: C₆H₃F₅N₂
MW: 198,10 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD03840200
CAS Number: 675602-89-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR