Benzyloxyacetaldehyde dimethyl acetal ≥97%

Supplier: Thermo Scientific

L08651.09 L08651.18
ALFAL08651.09EA 169 SGD
ALFAL08651.09 ALFAL08651.18
Benzyloxyacetaldehyde dimethyl acetal ≥97%
Benzyloxyacetaldehyde dimethyl acetal

Purity: 97% CAS No.: 127657-97-0 Molecular Formula: C6H5CH2OCH2CH(OCH3)2 Formula Weight: 196.24 Refractive Index: 1.4910 Boiling Point: 80-82°/0.5mm MDL No.: MFCD00082233

Formula: C₁₁H₁₆O₃
MW: 196,24 g/mol
Boiling Pt: 80…82 °C (0,5 mmHg)
Density: 1,035
Storage Temperature: Ambient
MDL Number: MFCD00082233
CAS Number: 127657-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR