(3-Chloropropyl)trimethylsilane ≥98%

Supplier: Thermo Scientific
Warning

L04458.06 L04458.14
ALFAL04458.06EA 0 SGD
ALFAL04458.06 ALFAL04458.14
(3-Chloropropyl)trimethylsilane ≥98%
(3-Chloropropyl)trimethylsilane

Purity: 98% CAS No.: 2344-83-4 Molecular Formula: ClCH2CH2CH2Si(CH3)3 Formula Weight: 150.73 Flash Point: 49°(120°F) Refractive Index: 1.4320 Boiling Point: 150-152° Beilstein Reference: 1732625 UN No.: 1993 MDL No.: MFCD00045299

Formula: C₆H₁₅ClSi
MW: 150,73 g/mol
Boiling Pt: 150…152 °C
Density: 0,879
Flash Pt: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045299
CAS Number: 2344-83-4
EINECS: 219-059-0
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR