Benzamidoxime 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N'-Hydroxybenzimidamide, Benzamide oxime
H26068.06
H26068.03
ALFAH26068.06EA
0
SGD
ALFAH26068.06
ALFAH26068.03
Benzamidoxime 95%
Benzamidoxime
Purity: 95%
CAS No.: 613-92-3
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 72-76°
MDL No.: MFCD00031485
Formula:
C₇H₈N₂O MW: 136,15 g/mol Melting Pt: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031485 CAS Number: 613-92-3 EINECS: 210-361-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...