Barium isopropoxide ≥99.5% (metals basis)
Supplier: Thermo Scientific
Synonyms:
Barium i-propoxide
41271.03
41271.06
ALFA41271.03EA
0
SGD
ALFA41271.03
ALFA41271.06
Barium isopropoxide ≥99.5% (metals basis)
Barium isopropoxide
Purity: 99.5%
CAS No.: 24363-37-9
Molecular Formula: C6H14BaO2
Formula Weight: 255.52
Physical Form: Powder/Lump
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: ca 200° dec.
UN No.: 3205
MDL No.: MFCD00050486
Formula:
C₆H₁₄BaO₂ MW: 255,501 g/mol Melting Pt: ∼200 °C (dec.) |
MDL Number:
MFCD00050486 CAS Number: 24363-37-9 UN: 3205 ADR: 4.2,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...