4-Methoxy-3-methylphenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Methoxy-3-methylbenzeneboronic acid
L19821.03
L19821.06
ALFAL19821.03EA
57.6
SGD
ALFAL19821.03
ALFAL19821.06
4-Methoxy-3-methylphenylboronic acid ≥97%
4-Methoxy-3-methylphenylboronic acid
Purity: 98+%
CAS No.: 175883-62-2
Molecular Formula: C8H11BO3
Formula Weight: 165.99
Melting Point: ca 215°
Beilstein Reference: 7475212
Formula:
C₈H₁₁BO₃ MW: 165,99 g/mol Melting Pt: ∼ 215 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179464 CAS Number: 175883-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...