1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester 95%

Supplier: Thermo Scientific

Synonyms: 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

H27783.MD H27783.03
ALFAH27783.MDEA 0 SGD
ALFAH27783.MD ALFAH27783.03
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester 95%
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester

Purity: 95% CAS No.: 1072944-26-3 Molecular Formula: C13H21BN2O3 Formula Weight: 264.13 Melting Point: 84-91° MDL No.: MFCD09037503

Formula: C₁₃H₂₁BN₂O₃
MW: 264,13 g/mol
Melting Pt: 84…91 °C
Storage Temperature: Ambient
MDL Number: MFCD09037503
CAS Number: 1072944-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR