4-Benzyloxy-2,6-dimethoxybenzaldehyde polymer-supported 1,0-1,5 mmol/g on Merrifield resin
Supplier: Thermo Scientific
L19754.03
L19754.14
ALFAL19754.03EA
0
SGD
ALFAL19754.03
ALFAL19754.14
4-Benzyloxy-2,6-dimethoxybenzaldehyde polymer-supported 1,0-1,5 mmol/g on Merrifield resin
4-Benzyloxy-2,6-dimethoxybenzaldehyde on resin
MDL No.: MFCD02683435
Formula:
C₁₆H₁₆O₄ Storage Temperature: Ambient |
CAS Number:
679428-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...