β-D-Allose ≥98%
Supplier: Thermo Scientific
Synonyms:
(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol, β-D-Allopyranose, (2R,3R,4R,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
A16810.MC
A16810.ME
ALFAA16810.MCEA
0
SGD
ALFAA16810.MC
ALFAA16810.ME
β-D-Allose ≥98%
β-D-Allose
Purity: 98%
CAS No.: 2595-97-3
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 143-146°
Beilstein Reference: 1724623
Merck Reference: 14,280
MDL No.: MFCD00063268
Formula:
C₆H₁₂O₆ MW: 180,16 g/mol Melting Pt: 143…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063268 CAS Number: 7283-09-2 EINECS: 219-994-4 Merck Index: 14,280 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...