5-Amino-4-cyano-1-phenyl-1H-pyrazole 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

B20679.03 B20679.06
ALFAB20679.03EA 0 SGD
ALFAB20679.03 ALFAB20679.06
5-Amino-4-cyano-1-phenyl-1H-pyrazole 98%
5-Amino-4-cyano-1-phenyl-1H-pyrazole

Purity: 98% CAS No.: 5334-43-0 Molecular Formula: C10H8N4 Formula Weight: 184.20 Melting Point: ca 138° dec. Beilstein Reference: 161503 UN No.: 3439 MDL No.: MFCD00020730

Formula: C₁₀H₈N₄
MW: 184.20 g/mol
Melting Pt: ∼ 138 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00020730
CAS Number: 5334-43-0
EINECS: 226-253-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR