3,5-Dichloro-4-hydroxybenzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

L20174.03 L20174.06
ALFAL20174.03EA 0 SGD
ALFAL20174.03 ALFAL20174.06
3,5-Dichloro-4-hydroxybenzenesulphonyl chloride ≥97%
3,5-Dichloro-4-hydroxybenzenesulphonyl chloride

Purity: 97% CAS No.: 13432-81-0 Molecular Formula: C6H3Cl3O3S Formula Weight: 261.51 Melting Point: 118-119° Beilstein Reference: 2806977 UN No.: 3261 MDL No.: MFCD00171551

Formula: C₆H₃Cl₃O₃S
MW: 261,51 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00171551
CAS Number: 13432-81-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR