3-(Trifluoromethoxy)salicylaldehyde ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-3-(trifluoromethoxy)benzaldehyde

H26147.MD H26147.03
ALFAH26147.MDEA 0 SGD
ALFAH26147.MD ALFAH26147.03
3-(Trifluoromethoxy)salicylaldehyde ≥99%
3-(Trifluoromethoxy)salicylaldehyde

Purity: 99% CAS No.: 497959-31-6 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Melting Point: 58-60° MDL No.: MFCD03788527

Formula: C₈H₅F₃O₃
MW: 206,12 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03788527
CAS Number: 497959-31-6
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR