2-Methoxy-6-(methylamino)pyridine 98%

Supplier: Thermo Scientific
Warning

L20048.06 L20048.14
ALFAL20048.06EA 0 SGD
ALFAL20048.06 ALFAL20048.14
2-Methoxy-6-(methylamino)pyridine 98%
2-Methoxy-6-(methylamino)pyridine

Purity: 98% CAS No.: 88569-83-9 Molecular Formula: C7H10N2O Formula Weight: 138.17 Refractive Index: 1.5595

Formula: C₇H₁₀N₂O
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191557
CAS Number: 88569-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR