6-Fluoro-2,3-dimethoxybenzaldehyde ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3-Dimethoxy-6-fluorobenzaldehyde
H51877.MD
H51877.03
ALFAH51877.MDEA
192
SGD
ALFAH51877.MD
ALFAH51877.03
6-Fluoro-2,3-dimethoxybenzaldehyde ≥97%
6-Fluoro-2,3-dimethoxybenzaldehyde
Purity: 97%
Molecular Formula: C9H9FO3
Formula Weight: 184.16
Melting Point: 45-47°
MDL No.: MFCD11110608
Formula:
C₉H₉FO₃ MW: 184,16 g/mol Melting Pt: 45…47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11110608 CAS Number: 457628-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...