4-Methyl-1-pentenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: (4-Methylpent-1-en-1-yl)boronic acid

H27655.MD H27655.03
ALFAH27655.MDEA 0 SGD
ALFAH27655.MD ALFAH27655.03
4-Methyl-1-pentenylboronic acid ≥97%
4-Methyl-1-pentenylboronic acid

Purity: 97% CAS No.: 214907-33-2 Molecular Formula: C6H13BO2 Formula Weight: 127.98 Melting Point: 99-102° MDL No.: MFCD01074680

Formula: C₆H₁₃BO₂
MW: 127,98 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD01074680
CAS Number: 214907-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR