4-(Methylsulphonyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid
L17720.03
L17720.06
L17720.MD
ALFAL17720.03EA
212
SGD
ALFAL17720.03
ALFAL17720.06
ALFAL17720.MD
4-(Methylsulphonyl)phenylboronic acid ≥98%
4-(Methylsulphonyl)phenylboronic acid
Purity: 98%
CAS No.: 149104-88-1
Molecular Formula: C7H9BO4S
Formula Weight: 200.02
Melting Point: ca 200°
MDL No.: MFCD01630820
Formula:
C₇H₉BO₄S MW: 200,02 g/mol Melting Pt: ∼ 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD01630820 CAS Number: 149104-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...