4-Hydroxy-4'-iodobiphenyl ≥98%
Supplier: Thermo Scientific
|
Warning
|
B22115.06
B22115.14
B22115.22
ALFAB22115.06EA
138
SGD
ALFAB22115.06
ALFAB22115.14
ALFAB22115.22
4-Hydroxy-4'-iodobiphenyl ≥98%
4-Hydroxy-4'-iodobiphenyl
Purity: 98+%
CAS No.: 29558-78-9
Molecular Formula: C12H9IO
Formula Weight: 296.11
Melting Point: 117-119°
MDL No.: MFCD02258843
Formula:
C₁₂H₉IO MW: 296,11 g/mol Melting Pt: 196…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258843 CAS Number: 29558-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...