1-Naphthyl boronic acid neopentyl glycol ester ≥98%

Supplier: Thermo Scientific

Synonyms: 1-Naphthaleneboronic acid neopentyl glycol ester, 5,5-Dimethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborinane

L17612.03 L17612.06
ALFAL17612.03EA 0 SGD
ALFAL17612.03 ALFAL17612.06
1-Naphthyl boronic acid neopentyl glycol ester ≥98%
1-Naphthyl boronic acid neopentyl glycol ester

Purity: 98% CAS No.: 22871-77-8 Molecular Formula: C15H17BO2 Formula Weight: 240.11 Melting Point: 67-69° Beilstein Reference: 3053756

Formula: C₁₅H₁₇BO₂
MW: 240,11 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01318167
CAS Number: 22871-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR