3-Bromo-4-fluorobiphenyl ≥98%
Supplier: Thermo Scientific
Synonyms:
3-Bromo-4-fluoro-1,1'-biphenyl
B25316.06
B25316.14
ALFAB25316.06EA
0
SGD
ALFAB25316.06
ALFAB25316.14
3-Bromo-4-fluorobiphenyl ≥98%
3-Bromo-4-fluorobiphenyl
Purity: 98%
CAS No.: 306935-88-6
Molecular Formula: C12H8BrF
Formula Weight: 251.10
UN No.: 3151
MDL No.: MFCD01571091
Formula:
C₁₂H₈BrF MW: 251,10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01571091 CAS Number: 306935-88-6 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...