(3,5-Difluorophenyl)methanesulphonyl chloride 95%

Supplier: Thermo Scientific
Danger

H34210.MD H34210.03
ALFAH34210.MDEA 0 SGD
ALFAH34210.MD ALFAH34210.03
(3,5-Difluorophenyl)methanesulphonyl chloride 95%
(3,5-Difluorophenyl)methanesulphonyl chloride
Formula: C₇H₅ClF₂O₂S
MW: 226,63 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Freezer
MDL Number: MFCD09743349
CAS Number: 163295-74-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR