2-(Methylsulphonyl)benzoic acid ≥98%

Supplier: Thermo Scientific
Warning

H51018.06 H51018.03
ALFAH51018.06EA 519 SGD
ALFAH51018.06 ALFAH51018.03
2-(Methylsulphonyl)benzoic acid ≥98%
2-(Methylsulphonyl)benzoic acid

Purity: 98% CAS No.: 33963-55-2 Molecular Formula: C8H8O4S Formula Weight: 200.21 Melting Point: 136-141° MDL No.: MFCD00216492

Formula: C₈H₈O₄S
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00216492
CAS Number: 33963-55-2
EINECS: 223-765-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR