Benzhydryl isothiocyanate ≥97%

Supplier: Thermo Scientific
Danger

L19249.03 L19249.06
ALFAL19249.03EA 132 SGD
ALFAL19249.03 ALFAL19249.06
Benzhydryl isothiocyanate ≥97%
Benzhydryl isothiocyanate

Purity: 97% CAS No.: 3550-21-8 Molecular Formula: C14H11NS Formula Weight: 225.31 Melting Point: 58-60° Boiling Point: 160-165°/4mm UN No.: 2811 MDL No.: MFCD00046815

Formula: C₁₄H₁₁NS
MW: 225,31 g/mol
Boiling Pt: 160…165 °C (4 mmHg)
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00046815
CAS Number: 3550-21-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR