Ethyl-2,6-dimethylbenzoate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Ethyl 2,6-dimethylbenzoate

L07381.03 L07381.06
ALFAL07381.03EA 0 SGD
ALFAL07381.03 ALFAL07381.06
Ethyl-2,6-dimethylbenzoate ≥98%
Ethyl 2,6-dimethylbenzoate

Purity: 98% CAS No.: 36596-67-5 Molecular Formula: C11H14O2 Formula Weight: 178.23 Refractive Index: 1.4980 Boiling Point: 78-80°/0.9mm MDL No.: MFCD00051785

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 78…80 °C (0.9 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051785
CAS Number: 36596-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR