Erlenmeyer flasks with screw cap, capacity scale, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
26505-250 26505-125 26505-500
KIML26505-250EA 1160 SGD
KIML26505-250 KIML26505-125 KIML26505-500
Erlenmeyer flasks with screw cap, capacity scale, KIMAX®, KIMBLE®
Flasks Erlenmeyer Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Included are long caps with cemented-in white rubber liners, not attached
  • All flasks have scales to indicate approximate volumes
Useful for mixing and storing culture media.
Certifications: Designed from ASTM specification E1404, Type III requirements, glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.