Erlenmeyer flasks, wide neck, heavy-duty rim, PYREX®
Supplier: Corning
5100-125 5100-1L 5100-250 5100-2L 5100-500
CORN5100-125EA 83.7 SGD
CORN5100-125 CORN5100-1L CORN5100-250 CORN5100-2L CORN5100-500
Erlenmeyer flasks, wide neck, heavy-duty rim, PYREX®
Flasks Erlenmeyer Flasks
Heavy-duty rim flasks are especially recommended for use as titration flasks.
  • White enamel graduation to show the approximate capacity
  • With extra large marking spot
  • Good thermal and chemical resistance
Designed for long life and maximum safety, these flasks have a wide mouth, as indicated by stopper openings.
Order Now
SPECIFICATIONS
Environmentally Preferable