Chromatographic reagent atomisers, PYREX®
Supplier: Corning
2153-125 2153-250
CORN2153-125EA 255 SGD
CORN2153-125 CORN2153-250
Chromatographic reagent atomisers, PYREX®
Atomisers
Made from Type I Class A - low-expansion Borosilicate glass, per ASTM E438 (standard specification for glasses in laboratory apparatus).
They have a full length 24/40 ST joint between the reservoir and spraying device.