Accessories for tall form measuring cylinders, class A, borosilicate glass, PYREX®
Supplier: Corning
3066-100 3066-1L 3066-25 3066-250 3066-50 3066-500
CORN3066-100EA 5.3 SGD
CORN3066-100 CORN3066-1L CORN3066-25 CORN3066-250 CORN3066-50 CORN3066-500
Accessories for tall form measuring cylinders, class A, borosilicate glass, PYREX®
Cylinders Measuring Cylinders
Polyethylene shock-absorbing rings can reduce breakage of glass cylinders. For best results, slide the ring toward the top of the cylinder.