Flat bottom culture tubes, rimless, PYREX®
Supplier: Corning
9850-25 9850-25X 9850-25XX
CORN9850-25EA 204 SGD
CORN9850-25 CORN9850-25X CORN9850-25XX
Flat bottom culture tubes, rimless, PYREX®
Tubes Culture Tubes
Flat-bottom culture tubes are ideal for plant tissue and general culture work.
Two-position polypropylene closures are sold separately. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimises contamination.
The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage.
Ordering information: Tubes and closures sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable