Tissue grinder, Potter-Elvehjem, PTFE, PYREX®
Supplier: Corning
7725T-17 7725T-3 7725T-45 7725T-5 7725T-5X 7725T-8 7725T-1
CORN7725T-17EA 227 SGD
CORN7725T-17 CORN7725T-3 CORN7725T-45 CORN7725T-5 CORN7725T-5X CORN7725T-8 CORN7725T-1
Tissue grinder, Potter-Elvehjem, PTFE, PYREX®
Homogenisers Tissue Grinders
Homogenisers designed for preparing small samples for pathological and biochemical studies.
Glass pestle has fluoropolymer resin contact surface threaded onto a stainless steel shaft. Mortar has a tooled mouth and is not ground. Sample and buffer are forced through cylindrical portion of mortar as pestle is rotated downward.
Caution: Caution: If used with motor-driven device, attachment must contain some form of friction release clutch to prevent connective tissue or muscle from damaging apparatus. User should wear protective gloves of sufficient thickness in case glass components fail.
Order Now
SPECIFICATIONS
Environmentally Preferable