Jars, cylindrical, PYREX®
Supplier: Corning
6942-17L 6942-27L 6942-32L 6942-7L 6942-9L
CORN6942-17LEA 938 SGD
CORN6942-17L CORN6942-27L CORN6942-32L CORN6942-7L CORN6942-9L
Jars, cylindrical, PYREX®
Jars General Use Jars
These PYREX® jars are mold-blown with substantial walls for mechanical strength.
  • Open end is ground flat
These jars can also be used as animal jars.
Caution: Do not use with heat, vacuum, or pressure applications.