Separating funnels, Squibb, with PTFE valve and screw cap, PYREX®
Supplier: Corning
6305-1L 6305-250 6305-2L 6305-2LFO 6305-4L 6305-500 6305-500FO 6305-60 6305-60FO 6305-6LFO
CORN6305-1LEA 345 SGD
CORN6305-1L CORN6305-250 CORN6305-2L CORN6305-2LFO CORN6305-4L CORN6305-500 CORN6305-500FO CORN6305-60 CORN6305-60FO CORN6305-6LFO
Separating funnels, Squibb, with PTFE valve and screw cap, PYREX®
Funnels Separating Funnels
Borosilicate glass funnels feature an unbreakable PTFE valve and drip tip and have a screw thread closure at the top. Supplied with a PTFE valve assembly and GL screw cap with PTFE-faced silicone liner. No stopper.
Replacement funnels do not include a PTFE valve and drip tip or cap.
Ordering information: For replacement valve bottoms or caps, contact your Avantor representative.
Order Now
SPECIFICATIONS
Environmentally Preferable