Short neck boiling flask, round bottom, [ST] joint, PYREX®
Supplier: Corning
4320-100 4320-12L 4320-1L 4320-250 4320-2L 4320-300 4320-50 4320-500 4320-5L 4320A-50 4320B-100 4320B-25 4320B-250 4320B-50 4320B-500 4320C-100 4320C-12L 4320C-1L 4320C-250 4320C-2L 4320C-3L 4320C-50 4320C-500 4320C-5L 4321-100 4321-1L 4321-250 4321-500 4321A-100 4321A-25 4321A-250 4321A-50 4321A-500
CORN4320-100EA 56.5 SGD
CORN4320-100 CORN4320-12L CORN4320-1L CORN4320-250 CORN4320-2L CORN4320-300 CORN4320-50 CORN4320-500 CORN4320-5L CORN4320A-50 CORN4320B-100 CORN4320B-25 CORN4320B-250 CORN4320B-50 CORN4320B-500 CORN4320C-100 CORN4320C-12L CORN4320C-1L CORN4320C-250 CORN4320C-2L CORN4320C-3L CORN4320C-50 CORN4320C-500 CORN4320C-5L CORN4321-100 CORN4321-1L CORN4321-250 CORN4321-500 CORN4321A-100 CORN4321A-25 CORN4321A-250 CORN4321A-50 CORN4321A-500
Short neck boiling flask, round bottom, [ST] joint, PYREX®
Flasks Round Bottom Flasks
Boiling flasks with full length [ST] outer joints are manufactured from borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With full length NS outer joints. Normal working temperature is 230 °C (446 °F). Heavy wall flasks feature walls that are 25 to 30% thicker than standard walls, for increased mechanical strength.
Certifications: Flasks reference ASTM E123.