Solvent still head, PYREX®
Supplier: Corning
3575-250 3575-500
CORN3575-250EA 677 SGD
CORN3575-250 CORN3575-500
Solvent still head, PYREX®
Distillation Components Distillation Heads
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Solvent still head for maintaining oxygen-free solvents. Head has sufficient storage space to allow solvent removal by syringe or drainage through 2 mm T-bore fluoropolymer resin stopcock. It has a NS outer joint at top, NS inner joint at bottom, and NS 14/20 inner joint on the drainage arm.