Kuderna-Danish concentrator apparatus, with keck clamps, PYREX®
Supplier: Corning
2157-1L 2157-500
CORN2157-1LEA 326 SGD
CORN2157-1L CORN2157-500
Kuderna-Danish concentrator apparatus, with keck clamps, PYREX®
Concentrators
The complete apparatus consists of a 3-ball Snyder column with [ST] 24/40 joints, flask with [ST] 24/40 outer upper joint and [ST] 19/22 inner lower joint, and graduated tube with [ST] 19/22 outer joint. Tubes are graduated from 0 to 2 ml in 0,1 ml intervals and from 3 ml to capacity in 1,0 ml intervals. All columns have venting dimples. Concentrators are supplied with Keck clamps rather than spring hooks.
Ordering information: The 500 ml concentrator is supplied with a 10 ml tube. The 1000 ml concentrator comes with a 25 ml tube.