Funnels, short stem, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
28950-75 28950-90 28950-100 28950-25 28950-45
KIML28950-75EA 409 SGD
KIML28950-75 KIML28950-90 KIML28950-100 KIML28950-25 KIML28950-45
Funnels, short stem, KIMAX®, KIMBLE®
Funnels Filtration Funnels Filter Funnels
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Funnels have a high resistance to chemical attack and mechanical and thermal shock
  • The KIMAX® 58° funnel promotes faster, more effective filtering because the cone is suspended by its uppermost edge, leaving most of the conical area for filtration
Constructed with a strong, molded rim, heavy uniform walls, and a fire-polished stem for a long service life.
Certifications: Designed from ASTM Specification E1095, Type I, Class A requirements. Glass conforming to USP Type I and ASTM E43More Product Information