Fritted Hirsch funnels, KONTES®, KIMBLE®
Supplier: KIMBLEKONTES
955250-3523 955250-5023 955250-5026 955250-5044
KIML955250-3523EA 116 SGD
KIML955250-3523 KIML955250-5023 KIML955250-5026 KIML955250-5044
Fritted Hirsch funnels, KONTES®, KIMBLE®
Funnels Filtration Funnels Filter Funnels
Made of borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Funnels feature a conical shape that directs filtrate through a reduced-area frit to concentrate crystal residue for rewashing.
Order Now
SPECIFICATIONS
Environmentally Preferable