Screw caps with PTFE faced rubber Liner, KIMBLE®
Supplier: KIMBLEKONTES
73802-13425 45066C-13 73802-22400 73802-8425
KIML73802-13425EA 251 SGD
KIML73802-13425 KIML45066C-13 KIML73802-22400 KIML73802-8425
Screw caps with PTFE faced rubber Liner, KIMBLE®
Closures Test/Sample Tube Closures
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Certifications: It meet ASTM specification E982, Class B requirements.