Pipette fillers, Azlon®
Supplier: SciLabware
PWY144 PWY145
AZLOPWY144EA 30.2 SGD
AZLOPWY144 AZLOPWY145
Pipette fillers, Azlon®
Pipettes Pipette Bulbs
Natural rubber pipette fillers for pipettes up to 100 ml.
  • Pipette fi ller works via a series of glass ball valves for air release, suction and delivery
  • PWY145 is suitable for single handed operation