Blood typing tubes, pre-labeled, KIMBLE®
Supplier: KIMBLEKONTES
60A10BZ1 60A10BZ2 60A10BZ3 60A10BZ4 60A10BZ5 60A10BZ6 60B12BZ2 60B12BZ3 60B12BZ4 60B12BZ5 60B12BZ6
KIML60A10BZ1EA 264 SGD
KIML60A10BZ1 KIML60A10BZ2 KIML60A10BZ3 KIML60A10BZ4 KIML60A10BZ5 KIML60A10BZ6 KIML60B12BZ2 KIML60B12BZ3 KIML60B12BZ4 KIML60B12BZ5 KIML60B12BZ6
Blood typing tubes, pre-labeled, KIMBLE®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
  • Pre-labelled with blood type
  • Convenient package quantities
  • Permanent yellow labels placed to allow an unobstructed view of cell suspension
These culture tubes are manufactured with a fused ceramic paint to form writing labels that will not separate from the borosilicate glass tube. Labels measure 1,9W×3,5L cm.
Order Now
SPECIFICATIONS
Environmentally Preferable