Culture tubes, disposable, with marking spot, KIMBLE®
Supplier: KIMBLEKONTES
60E16BZW 60A10BZW 60B12BZW
KIML60E16BZWEA 512 SGD
KIML60E16BZW KIML60A10BZW KIML60B12BZW
Culture tubes, disposable, with marking spot, KIMBLE®
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
  • Permanent ceramic labels are located ³/₄" from the open end
  • All tubes have sturdy, uniform bottoms and consistent lengths
  • Borosilicate glass tubes have excellent chemical resistance against sodium leaching
These Chase brand Mark-M™ glass culture tubes feature a permanent vertical ceramic label, fused to the glass, which is easily written on with marker or pencil.
Order Now
SPECIFICATIONS
Environmentally Preferable