Tissue grinders with PTFE pestles, Potter-Elvehjem, KONTES®, KIMBLE®
Supplier: KIMBLEKONTES
886000-0019 886000-0022 886000-0021
KIML886000-0019EA 375 SGD
KIML886000-0019 KIML886000-0022 KIML886000-0021
Tissue grinders with PTFE pestles, Potter-Elvehjem, KONTES®, KIMBLE®
Homogenisers Tissue Grinders
For homogenisation of tissue samples. Grinders have a PTFE pestle and an ungrounded borosilicate glass tube.
Sizes 21 to 24 have radial serrations on the lower portion of the pestle to deliver the homogenate into the cylindrical portion of the tube. Tubes and pestles are interchangeable, with a uniform 0,10 to 0,15 mm clearance.
Ordering information: Special clearance pestles are available upon request; contact your Avantor sales representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable